Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100713
Description
CCR6 Polyclonal specifically detects CCR6 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR6 | |
Polyclonal | |
Unconjugated | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:250, Flow Cytometry 2-6 ul/10^6 cells in 100ul, ELISA 1:37500, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 | |
P51684 | |
CCR6 | |
Synthetic peptide: YFVSVNTSYYSVDSEMLLCSLQEVRQFSR, corresponding to amino acids 18-46 of Human CCR6. | |
0.1 mg | |
Chemokines and Cytokines | |
1235 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction