Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
$464.00
Specifications
Antigen | CCR6 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:250, Flow Cytometry 2-6 ul/10^6 cells in 100ul, ELISA 1:37500, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
CCR6 Polyclonal specifically detects CCR6 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR6 | |
Western Blot 1:250, Flow Cytometry 2-6 ul/10^6 cells in 100ul, ELISA 1:37500, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 | |
Polyclonal | |
Goat | |
Chemokines and Cytokines | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
CCR6 | |
IgG | |
Affinity Purified |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
P51684 | |
1235 | |
Synthetic peptide: YFVSVNTSYYSVDSEMLLCSLQEVRQFSR, corresponding to amino acids 18-46 of Human CCR6. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title