Learn More
CCR7 Antibody (3D12) - BSA Free, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP1433320.1MG
 View more versions of this product
Catalog No. NBP14333201
Only null left
Add to Cart
Description
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CCR7
Monoclonal
0.5 mg/mL
Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P32248
CCR7
The immunogen for this antibody was CCR7.
0.1 mg
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1236
Store at 4C. Do not freeze.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
3D12
Unconjugated
PBS, pH 7.2 with 0.09% Sodium Azide
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Rat
Affinity Purified
RUO
Primary
Human
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only