Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 Antibody (3D12) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen CCR7
Clone 3D12
Concentration 0.5 mg/mL
Dilution Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018534
SDP
View Documents
Novus Biologicals
NBP14333225UG
25 μg
Each for $206.50
Only null left
Add to Cart
 
NBP14333201
SDP
View Documents
Novus Biologicals
NBP1433320.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CCR7
0.5 mg/mL
Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry
Unconjugated
RUO
Human
P32248
1236
The immunogen for this antibody was CCR7.
Primary
Store at 4C. Do not freeze.
3D12
Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Rat
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
PBS, pH 7.2 with 0.09% Sodium Azide
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
CCR7
IgG2a κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.