Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 Antibody (4B7B8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61680 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61680 Supplier Novus Biologicals Supplier No. NBP261680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen CCR7
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 4B7B8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100- 1:500, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Gene Symbols CCR7
Host Species Mouse
Immunogen Purified recombinant fragment of human CD197 (AA: extra mix) expressed in E. Coli.
Molecular Weight of Antigen 42.9 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1236
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.