Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB199950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199950 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199950 Supplier Novus Biologicals Supplier No. NBP241251F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCR7 Polyclonal antibody specifically detects CCR7 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CCR7
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the center of human CCR7. The immunogen is located within amino acids 180 - 230 of CCR7. Amino Acid Squence: LYSDLQRSSSEQAMR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1236
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.