Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR8 Antibody (2C10), Novus Biologicals™
SDP

Catalog No. NB297600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297600 50 μL
NB297601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297600 Supplier Novus Biologicals Supplier No. NBP32668850UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

CCR8 Monoclonal antibody specifically detects CCR8 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CCR8
Applications Flow Cytometry
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Dilution Flow Cytometry
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1
Host Species Human
Immunogen Recombinant Human CCR8 protein [UniProt P51685]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1237
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.