Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR8 Antibody, Novus Biologicals™
SDP

Catalog No. NB017801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017801 0.025 mg
NB100709 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017801 Supplier Novus Biologicals Supplier No. NB1007090.025MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

CCR8 Polyclonal specifically detects CCR8 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen CCR8
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200
Formulation 10mM KHPO4 and 0.14M NaCl, BSA with 0.1% Sodium Azide
Gene Accession No. P56484
Gene Alias CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1
Gene Symbols CCR8
Host Species Goat
Immunogen Synthetic peptide: PNVTMTDYYPDFFTAPCDAEFLLRGSM, corresponding to amino acids 7-33 of Mouse CCR8.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1237
Test Specificity This recognizes CCR8 (50 kDa). The peptide sequence is < 50 % identical to other mouse chemokine receptors in this region.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.