Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR8 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
$207.00 - $512.00
Specifications
Antigen | CCR8 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA |
Classification | Polyclonal |
Description
CCR8 Polyclonal specifically detects CCR8 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR8 | |
Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Goat | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
10mM KHPO4 and 0.14M NaCl, BSA with 0.1% Sodium Azide | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
CCR8 | |
IgG | |
Affinity Purified | |
This recognizes CCR8 (50 kDa). The peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA | |
Unconjugated | |
RUO | |
Mouse | |
P56484 | |
1237 | |
Synthetic peptide: PNVTMTDYYPDFFTAPCDAEFLLRGSM, corresponding to amino acids 7-33 of Mouse CCR8. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title