Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215767 Supplier Novus Biologicals Supplier No. NBP215767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCR8 Polyclonal specifically detects CCR8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CCR8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P51685
Gene Alias CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1
Gene Symbols CCR8
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CCR8. The exact sequence is proprietary.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1237
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.