Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR9 Antibody (112509), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB367524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367524 Supplier Novus Biologicals Supplier No. FAB179W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR9 Monoclonal antibody specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen CCR9
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 112509
Conjugate DyLight 650
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human CCR9, Met1-Leu369, Accession # P51686
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.