Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR9 Antibody (248621), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB314132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314132 Supplier Novus Biologicals Supplier No. FAB1791APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR9
Applications Flow Cytometry
Classification Monoclonal
Clone 248621
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Host Species Mouse
Immunogen Human CCR9 transfectants, Met1-Leu369, Accession # P51686
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10803
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.