Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR9 Antibody (9B1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB152789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152789 Supplier Novus Biologicals Supplier No. NBP312002AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR9 Monoclonal antibody specifically detects CCR9 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CCR9
Applications Flow Cytometry
Classification Monoclonal
Clone 9B1
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Host Species Rat
Immunogen This antibody was raised by immunizing rats with KLH-conjugated linear peptide corresponding to the N-terminus of mouse CCR9.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10803
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.