Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR9 Antibody (C9Mab-1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB158595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158595 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158595 Supplier Novus Biologicals Supplier No. NBP314623V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR9 Monoclonal antibody specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen CCR9
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone C9Mab-1
Conjugate DyLight 405
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Host Species Mouse
Immunogen CCR9 transfected CHO cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10803
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.