Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2/CRAM-A/B Antibody (BZ2E3), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB152287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152287 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152287 Supplier Novus Biologicals Supplier No. NBP311982C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen CCRL2/CRAM-A/B
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone BZ2E3
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor
Host Species Rabbit
Immunogen This antibody was raised by immunising Wistar Furth rats with the mCCRL2/CRAM-A/B peptide/KLH conjugate. The immunizing amino-terminal mCCRL2/CRAM-A/B peptide with the sequence NH2-MDNYTVAPDDEYDVLILDDYLDNSC-COOH, corresponding to residues 1-24 of mCCRL2/CRAM-A/B, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9034
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.