Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2/CRAM-A/B Antibody (MM0158-5A15), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP212137R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212137R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212137R Supplier Novus Biologicals Supplier No. NBP212137R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCRL2/CRAM-A/B Monoclonal specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CCRL2/CRAM-A/B
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0158-5A15
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor
Gene Symbols CCRL2
Host Species Mouse
Immunogen Human recombinant CCRL2/CRAM-A/B
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9034
Test Specificity Detects both human CRAM-Aand CRAM-B isoforms.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.