Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2/LCCR Antibody (498321), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB377867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377867 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377867 Supplier Novus Biologicals Supplier No. FAB5519M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCRL2/LCCR Monoclonal antibody specifically detects CCRL2/LCCR in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen CCRL2/LCCR
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 498321
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACKR5;C-C chemokine receptor-like 2;CKRX;CRAM;CRAM-A;CRAM-B;HCR
Host Species Rat
Immunogen RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR, Accession # O35457
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9034
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.