Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB084528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084528 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084528 Supplier Novus Biologicals Supplier No. NBP299266AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCT3 Polyclonal specifically detects CCT3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCT3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Cctg, chaperonin containing TCP1, subunit 3 (gamma), hTRiC5, TCP1 (t-complex-1) ring complex, polypeptide 5, TRIC5gamma subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CCT3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7203
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.