Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT8 Antibody (PK/13/72/8k) - BSA Free
SDP

Catalog No. NB10066596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10066596 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10066596 Supplier Novus Biologicals Supplier No. NB10066596

Rat Monoclonal Antibody

CCT8 Monoclonal specifically detects CCT8 in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CCT8
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone PK/13/72/8k
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500, Immunoprecipitation 1:10-1:500
Gene Accession No. P50990, Q3ZCI9
Gene Alias C21orf112, CCTQ, CCT-theta, chaperonin containing TCP1, subunit 8 (theta), chromosome 21 open reading frame 112, D21S246, KIAA0002, PRED71, T-complex protein 1 subunit theta, T-complex protein 1, theta subunit, 10Renal carcinoma antigen NY-REN-15, TCP-1-theta
Gene Symbols CCT8
Host Species Rat
Immunogen Made to CCT THETA
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10694
Test Specificity Recognises the theta polypeptide of the CCT chaperonin molecule complex.
Target Species Human, Mouse, Bovine, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.