Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD-M6PR Mouse anti-Human, Alexa Fluor 488, Clone: 2B4, Novus Biologicals™
SDP

Catalog No. NBP242667AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242667AG 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242667AG Supplier Novus Biologicals Supplier No. NBP242667AF488

Mouse Monoclonal Antibody

CD-M6PR Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.

Specifications

Antigen CD-M6PR
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B4
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR
Gene Symbols M6PR
Host Species Mouse
Immunogen Recombinant human M6PR (27-185aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4074
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.