Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1.1 antigen Antibody (CB3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N128362PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N128362PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N128362PEC7 Supplier Novus Biologicals Supplier No. NBP128362PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1.1 antigen Monoclonal specifically detects CD1.1 antigen in Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen CD1.1 antigen
Applications Flow Cytometry
Classification Monoclonal
Clone CB3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a
Gene Symbols CD1A
Host Species Mouse
Immunogen Lymphocytes from the bursa of Fabricius of outbred chickens
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Dendritic Cell Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 909.0
Test Specificity Chicken CD1.1
Target Species Chicken
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.