Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1 Antibody (76-7-4), FITC, Novus Biologicals™
SDP

Catalog No. NBP128222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128222 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128222 Supplier Novus Biologicals Supplier No. NBP128222
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1 Monoclonal specifically detects CD1 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen CD1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 76-7-4
Concentration 0.5mg/mL
Conjugate FITC
Dilution Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Gene Alias CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a
Gene Symbols CD1A
Host Species Mouse
Immunogen Fresh dd miniature swine thymocytes
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 909
Test Specificity Porcine CD1
Target Species Pig
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.