Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD109 Antibody (W7C5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB002866 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002866 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002866 Supplier Novus Biologicals Supplier No. NBP275969IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD109 Monoclonal specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen CD109
Applications Flow Cytometry
Classification Monoclonal
Clone W7C5
Conjugate DyLight 755
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1
Gene Symbols CD109
Host Species Mouse
Immunogen WERI-RB-1 retinoblastoma cell line
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 135228
Test Specificity The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.