Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD112 (Nectin-2) Recombinant Rabbit Monoclonal Antibody (BLR071G)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | CD112 (Nectin-2) |
---|---|
Clone | BLR071G |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544333
|
Invitrogen™
MA544333 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544334
|
Invitrogen™
MA544334 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
CD112 (Nectin-2) | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 7.8-8.6 | |
Q92692 | |
5819 | |
Peptide representing a region between residue 50 and 100 of human Nectin cell adhesion molecule 2. | |
Antibody |
BLR071G | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
NECTIN2 | |
AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs | |
NECTIN2 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title