Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD114 (G-CSFR) Monoclonal Antibody (LMM741), PerCP-eFluor™ 710, eBioscience™
GREENER_CHOICE

Catalog No. 501130777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-0777 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-0777 Supplier Invitrogen™ Supplier No. 46114942
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This LMM741 monoclonal antibody reacts with human CD114, which is also known as granulocyte colony-stimulating factor receptor (G-CSFR). A type I hematopoietin cytokine receptor, CD114 is expressed on granulocytes such as neutrophils, myeloid cells, monocytes, and leukemic cells. This receptor is absent from lymphocytes, erythrocytes, and eosinophils. CD114 can also be detected on non-hematopoietic cells, including neurons, endothelial cells, and cardiomyocytes. Studies demonstrate that expression of CD114 can be downregulated by LPS, TNF, phorbol ester, and G-CSF itself. Binding of G-CSF binding induces homodimerization of the receptor that activates signaling transduction (e.g., the JAK/STAT and Erk1/2 pathways) to mediate cell proliferation, survival, and differentiation. Applications Reported: This LMM741 antibody has been reported for use in flow cytometric analysis. Applications Tested: This LMM741 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass...

CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD114 (G-CSFR)
Applications Flow Cytometry
Classification Monoclonal
Clone LMM741
Concentration 5 μL/Test
Conjugate PerCP-eFluor 710
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CSF3R
Gene Accession No. Q99062
Gene Alias CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7
Gene Symbols CSF3R
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.