Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-kit Antibody (351), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB317101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317101 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317101 Supplier Novus Biologicals Supplier No. NBP290037APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen CD117/c-kit
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 351
Conjugate APC
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD117/c-kit (Uniprot#: P10721-2; Met1-Thr516).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3815
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.