Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-kit Antibody, Novus Biologicals™
SDP

Catalog No. NB406488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406488 25 μL
NBP185593 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406488 Supplier Novus Biologicals Supplier No. NBP18559325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

CD117/c-kit Polyclonal specifically detects CD117/c-kit in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD117/c-kit
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Gene Symbols KIT
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:VGDEIRLLCTDPGFVKWTFEILDETNENKQNEWITEKAEATNTGKYTCTNKHGLSNSIYVFVRDPAKLFLVDRSLYGKEDNDTLVRCPLTDPEVTNYSLKGCQGKPLPKDLRFIPDPKAGIMIKSVKRAYHRLCLHCSVDQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3815
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.