Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-kit Antibody (OTI2B12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP271076IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271076IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271076IR Supplier Novus Biologicals Supplier No. NBP271076IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD117/c-kit
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2B12
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Gene Symbols KIT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 27-308 of human CD117/c-kit (NP_000213) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3815.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.