Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Novus Biologicals™ CD117/c-kit Antibody Pair [HRP]
 
    
    
    
        
            Shop All R&D Systems Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                15 Plates
                            
                        
                            
                                5 Plates
                            
                        
                    Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD117/c-kit.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| Assay Range | 23.4375 pg/mL | 
| Assay Sensitivity | 23.44-1500 pg/mL | 
| Assay | Sandwich ELISA | 
| Conjugate | HRP | 
| Product Type | Antibody Pairs | 
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD117/c-kit. | 
| Target Species | Human | 
| For Use With (Application) | Sandwich ELISA | 
| Gene Alias | CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | 
| Gene ID (Entrez) | 3815 | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            ![Novus Biologicals CD117/c-kit Antibody Pair [HRP] 15 Plates | Buy Online | Novus Biologicals™ | Fisher Scientific](https://assets.fishersci.com/TFS-Assets/CCG/product-images/default.jpg-250.jpg)