Learn More
CD117 Mouse anti-Human, PE, Clone: YB5.B8, BD
Mouse Monoclonal Antibody
$138.60 - $295.50
Specifications
Antigen | CD117 |
---|---|
Clone | YB5.B8 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The YB5.B8 monoclonal antibody specifically binds to CD117. CD117, also known as c-Kit, is a 145 kDa cell-surface glycoprotein with tyrosine kinase activity. CD117 is present on hematopoietic progenitor cell subsets, thymocytes, mast cells, hepatocytes and histiocytes. CD117 serves as a cytokine receptor for steel factor (SLF), also known as stem cell factor (SCF) or mast cell growth factor (MGF). The interaction of c-Kit and SLF is crucial to hematopoiesis, mast cell differentiation, melanogenesis, and germ cell development. The ability of the YB5.B8 antibody to block the binding of c-Kit ligand is still controversial.
Specifications
CD117 | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Human | |
Acute myelocytic leukemia blasts | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
YB5.B8 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
KIT; c-Kit; SCFR; PBT; Mast/stem cell growth factor receptor |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.