Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a/CD18 Antibody (hu1124 (Efalizumab)), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB275912A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275912A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275912A48 Supplier Novus Biologicals Supplier No. NBP275912AF488
Add to Cart
Add to Cart

Human Monoclonal Antibody

CD11a/CD18 Monoclonal specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CD11a/CD18
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone hu1124 (Efalizumab)
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Gene Symbols ITGAL
Host Species Human
Immunogen Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3683.0
Test Specificity Binds to human LFA-1.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.