Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a Mouse anti-Human, Rhesus, Cynomolgus, Baboon, Dog, Rabbit, R718, Clone: HI111, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567221
This item is not returnable.
View return policy
Description
CD11a Monoclonal antibody specifically detects CD11a in Human, Rhesus, Cynomolgus, Baboon, Dog, Rabbit samples. It is validated for Flow Cytometry
Specifications
| CD11a | |
| Monoclonal | |
| R718 | |
| LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
| IgG1 κ |
| Flow Cytometry | |
| HI111 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human, Rhesus Macaque, Cynomolgus, Baboon, Dog, Rabbit | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction