Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a Mouse anti-Human, Rhesus, Cynomolgus, Baboon, Dog, Rabbit, R718, Clone: HI111, BD Horizon™
Mouse Monoclonal Antibody
$144.00 - $303.50
Specifications
Antigen | CD11a |
---|---|
Clone | HI111 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
CD11a Monoclonal antibody specifically detects CD11a in Human, Rhesus, Cynomolgus, Baboon, Dog, Rabbit samples. It is validated for Flow CytometrySpecifications
CD11a | |
Flow Cytometry | |
R718 | |
Mouse | |
Human, Rhesus Macaque, Cynomolgus, Baboon, Dog, Rabbit | |
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
Primary |
HI111 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title