Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody (908649), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB350169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350169 Supplier Novus Biologicals Supplier No. FAB11241MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen CD11b
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 908649
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha M beta 2, Phe17-Asn1105 (Integrin alpha M), Gln24-Asn702 (Integrin beta 2), Accession # NP_032427 (Integrin alpha M) and P11835 (Integrin beta 2)
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.