Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB11089474F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11089474F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11089474F Supplier Novus Biologicals Supplier No. NB11089474F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD11b Polyclonal specifically detects CD11b in Human, Mouse, Rat, Bovine, C. elegans, Chinese Hamster, Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Product Image.

Specifications

Antigen CD11b
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P11215
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement
Gene Symbols ITGAM
Host Species Rabbit
Immunogen Rabbit Polyclonal CD11b Antibody was made to a synthetic peptide within residues 250-350 of the mouse CD11b protein. [Swiss-Prot# P05555]
Molecular Weight of Antigen 127.2 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Mouse, rat and human. We have feedback that this antibody does not work in human samples with Western blot. Immunogen has 100% homology to bovine and 88% homology with sheep.
Target Species Human, Mouse, Rat, Bovine, C. elegans, Chinese Hamster, Monkey, Rhesus Macaque
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.