Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody (ITGAM/271), Novus Biologicals™
SDP

Catalog No. NB253402200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253402200 200 μg
NB25340220 20 μg
NB253402100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253402200 Supplier Novus Biologicals Supplier No. NBP253402200UG

Mouse Monoclonal Antibody

CD11b Monoclonal antibody specifically detects CD11b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, SDS-Page.

Specifications

Antigen CD11b
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
Classification Monoclonal
Clone ITGAM/271
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Gene Symbols ITGAM
Host Species Mouse
Immunogen Recombinant human ITGAM protein
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.