Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody (M1/70.15) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013699 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013699 Supplier Novus Biologicals Supplier No. NBP280626
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD11b Monoclonal specifically detects CD11b in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen CD11b
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M1/70.15
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50. Use reported in scientific literature (PMID 28683563), Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported by customer review, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 28700779), Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature, CyTOF-ready
Formulation PBS
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Gene Symbols ITGAM
Host Species Rat
Immunogen Rat Monoclonal CD11b Antibody (M1/70.15) was made to T cell enriched splenocytes from B10 mice.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Mouse reactivity reported in a verified customer review. Rabbit reactivity reported in scientific literature (PMID: 28700779). Human reactivity reported in scientific literature (PMID: 28683563).
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.