Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody (M1/70.15), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N601327PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N601327PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N601327PEC5 Supplier Novus Biologicals Supplier No. NB6001327PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD11b Monoclonal specifically detects CD11b in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry.

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone M1/70.15
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement
Gene Symbols ITGAM
Host Species Rat
Immunogen Rat Monoclonal CD11b Antibody (M1/70.15) was made to T cell enriched splenocytes from B10 mice.
Molecular Weight of Antigen 127.2 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684.0
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.