Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody (OX-42), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB000986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000986 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000986 Supplier Novus Biologicals Supplier No. NB10065284PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11b Monoclonal specifically detects CD11b in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen CD11b
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone OX-42
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Gene Symbols ITGAM
Host Species Mouse
Immunogen Mouse Monoclonal CD11b Antibody (OX-42) was made from rat peritoneal macrophages.
Molecular Weight of Antigen 127.9 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity NB100-65284 recognizes the rat equivalent of human CD11b, the receptor for the iC3b component of complement. The antigen is expressed on most macrophages, including resident and activated peritoneal macrophages and Kupffer cells and around 35% of alveolar macrophages. This CD11b antibody also labels dendritic cells, granulocytes and microglial cells in the brain. Clone MRC OX-42 has been reported as being suitable for use on paraffin-embedded material following PLP fixation (periodate-lysine-paraformaldehyde). Functionally, clone OX-42 inhibits complement mediated rosettes.
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.