Learn More
Description
Specifications
Specifications
| Antigen | CD11b/c |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 |
| Gene Symbols | ITGAM |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an internal region (within residues 500-600) of the mouse CD11 (b/c) protein. [Swiss-Prot: P05555] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
