Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b/c Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11040766T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040766T 0.025 mL
NB11040766 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11040766T Supplier Novus Biologicals Supplier No. NB11040766SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 26 publications

CD11b/c Polyclonal specifically detects CD11b/c in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD11b/c
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Flow Cytometry 1:50 - 1:200, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 29467366)
Formulation PBS, 30% Glycerol with 0.02% Sodium Azide
Gene Accession No. P11215
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Gene Symbols ITGAM
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 500-600) of the mouse CD11 (b/c) protein. [Swiss-Prot: P05555]
Molecular Weight of Antigen 160 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Glia Markers, Immunology, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 25150592).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.