Learn More
Description
Specifications
Specifications
| Antigen | CD11b/c |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Flow Cytometry, Immunohistochemistry 5-10 μg/mL, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 5-10 μg/mL, Immunohistochemistry-Frozen |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P11215 |
| Gene Alias | antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 |
| Gene Symbols | ITGAM |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
