Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Antibody (118/A5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395539 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395539 Supplier Novus Biologicals Supplier No. NBP2501300.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen CD11c
Applications Immunohistochemistry
Classification Monoclonal
Clone 118/A5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Host Species Mouse
Immunogen Synthetic peptide conjugated to KLH corresponding to the C terminus of human CD11C: ANGQIAPENGTQTPSPPSEK
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3687
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.