Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (118/A5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | CD11c |
---|---|
Clone | 118/A5 |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:10 - 1:500 |
Applications | Immunohistochemistry |
Description
CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for ImmunohistochemistrySpecifications
CD11c | |
1 mg/mL | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Dendritic Cell Markers, Signal Transduction | |
PBS | |
3687 | |
IgG2b | |
Protein A purified |
118/A5 | |
Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
Synthetic peptide conjugated to KLH corresponding to the C terminus of human CD11C: ANGQIAPENGTQTPSPPSEK | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title