Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Antibody (AP-MAB0806) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB110978710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110978710 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110978710 Supplier Novus Biologicals Supplier No. NB110978710.1MG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody has been used in 12 publications

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation

CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen CD11c
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone AP-MAB0806
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:25-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:25-1:100, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9QXH4
Gene Alias 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Gene Symbols ITGAX
Host Species Armenian Hamster
Immunogen This CD11c antibody was produced from a hybridoma (mouse myeloma fused with spleen cells)from an Armenian Hamster immunized with mouse lymphocytes.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3687
Reconstitution Reconstitute with 0.4mL sterilized PBS to a final concentration of 0.5mg/mL.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.