Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Antibody (N418), FITC, Novus Biologicals™
SDP

Catalog No. NB015259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015259 Supplier Novus Biologicals Supplier No. NBP281066F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD11c
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone N418
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Gene Symbols ITGAX
Host Species Rabbit
Immunogen Hamster spleen cells that had been primed to mouse spleen dendritic cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3687
Test Specificity The antibody binds to mouse and human CD11c.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.