Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11C Armenian Hamster anti-Mouse, BB700, Clone: HL3, BD Horizon™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 566504
Description
CD11C Monoclonal antibody specifically detects CD11C in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD11C | |
Monoclonal | |
0.2 mg/mL | |
Armenian Hamster | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
HL3 | |
Brilliant Blue 700 (BB700) | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 λ2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction