Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11C Armenian Hamster anti-Mouse, BB700, Clone: HL3, BD Horizon™
Armenian Hamster Monoclonal Antibody
$140.60 - $382.00
Specifications
Antigen | CD11C |
---|---|
Clone | HL3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD11C Monoclonal antibody specifically detects CD11C in Mouse samples. It is validated for Flow Cytometry.Specifications
CD11C | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG1 λ2 | |
Refrigerate (2 to 8°C) |
HL3 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Armenian Hamster | |
Mouse | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title