Learn More
Description
Specifications
Specifications
| Antigen | CD11c |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | N418 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 |
| Gene Symbols | ITGAX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
