Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB007047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007047 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007047 Supplier Novus Biologicals Supplier No. NBP244249AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD133
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Gene Symbols PROM1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 300-400) [Uniprot: O54990]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.